missing translation for 'onlineSavingsMsg'
Learn More

6-methoxypyrid-2-ylamine, 97%, Thermo Scientific™

Product Code. 10789595
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10789595 1 g Amber Glass Bottle 1g
10386714 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10789595 Supplier Thermo Scientific Maybridge Supplier No. MO08464DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 17920-35-3
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD04972542
InChI Key DEUALFRBMNMGDS-UHFFFAOYSA-N
Synonym 2-amino-6-methoxypyridine, 6-methoxy-pyridin-2-ylamine, 6-methoxy-2-pyridinamine, 6-methoxypyrid-2-ylamine, 6-methoxy-2-pyridylamine, 2-amino-6-methoxy pyridine, 2-pyridinamine, 6-methoxy, methoxypyridin-6-amine, acmc-209efc, 6-methoxy-2-aminopyridin
PubChem CID 580053
IUPAC Name 6-methoxypyridin-2-amine
SMILES COC1=CC=CC(=N1)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 6-Methoxypyrid-2-ylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.