missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-1-octanol, TRC

Product Code. 30474636
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474636 500 mg 500mg
30475314 100 mg 100mg
30474250 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474636 Supplier Toronto Research Chemicals Supplier No. TRCM219480500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38514-05-5
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.25
IUPAC Name 6-methyloctan-1-ol
SMILES CCC(C)CCCCCO

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H20O/c1-3-9(2)7-5-4-6-8-10/h9-10H,3-8H2,1-2H3
Formula Weight 144.1514
Chemical Name or Material 6-Methyl-1-octanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.