missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-2-oxo-5-phenyl-1,2-dihydropyridine-3-carboxylic Acid, TRC

Product Code. 30336249
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30336249 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336249 Supplier Toronto Research Chemicals Supplier No. TRCB49646010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Methyl-2-oxo-5-phenyl-1,2-dihydropyridine-3-carboxylic Acid
CAS 10176-79-1
Molecular Formula C13H11NO3
Quantity 10 mg
InChI Formula InChI=1S/C13H11NO3/c1-8-10(9-5-3-2-4-6-9)7-11(13(16)17)12(15)14-8/h2-7H,1H3,(H,14,15)(H,16,17)
SMILES CC1=C(C=C(C(=O)O)C(=O)N1)c2ccccc2
IUPAC Name 6-methyl-2-oxo-5-phenyl-1H-pyridine-3-carboxylic acid
Molecular Weight (g/mol) 229.23
Formula Weight 229.0739
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.