missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methyl-2-pyridylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
Click to view available options
Quantity:
50 mL
Packaging:
ChemSeal™ bottles
Unit Size:
50mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Packaged under Argon
Specifications
Specifications
| CAS | 308795-98-4 |
| Color | Black to Yellow |
| Density | 0.996 g/mL |
| Packaging | ChemSeal™ bottles |
| Molecular Formula | C6H6BrNZn |
| MDL Number | MFCD01311462 |
| Quantity | 50 mL |
| UN Number | UN3399 |
| Sensitivity | Air sensitive |
| Synonym | zinc 2+ ion 6-methylpyridin-2-ide bromide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction