missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-3,5-heptadiene-2-one, 96%, Thermo Scientific™

Product Code. 15502323 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15502323 5 g 5g
15512323 25 g 25g
15522323 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15502323 Supplier Thermo Scientific Acros Supplier No. 453210050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1604-28-0
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.18
InChI Key KSKXSFZGARKWOW-GQCTYLIASA-N
Synonym 6-methyl-3,5-heptadien-2-one, 6-methylhepta-3,5-dien-2-one, methylheptadienone, unii-p7cmp2e76c, 3,5-heptadien-2-one, 6-methyl, 3e-6-methylhepta-3,5-dien-2-one, 3,5-heptadien-2-one, 6-methyl-, e, 6-methyl-trans-3,5-heptadien-2-one, p7cmp2e76c, fema no. 3363
PubChem CID 5370101
IUPAC Name (3E)-6-methylhepta-3,5-dien-2-one
SMILES CC(=CC=CC(=O)C)C

Specifications

Density 0.8930g/mL
Boiling Point 168.0°C
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Linear Formula (CH3)2C=CHCH=CHCOCH3
Quantity 5 g
Specific Gravity 0.893
Formula Weight 124.18
Percent Purity 96%
Chemical Name or Material 6-Methyl-3,5-heptadiene-2-one
Show More Show Less
compliance-icons
Product Identifier
  • 6-Methyl-3,5-heptadien-2-one
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.