missing translation for 'onlineSavingsMsg'
Learn More

6-Methylbenzoxazole, 99.5+%, Thermo Scientific Chemicals

Product Code. 10177933 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10177933 10 g Glass bottle 10g
10104550 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10177933 Supplier Thermo Scientific Supplier No. 293110100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10531-80-3
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00216937
InChI Key SZWNDAUMBWLYOQ-UHFFFAOYSA-N
Synonym 6-methylbenzo d oxazole, 6-methylbenzoxazole, 6-methyl-benzooxazole, benzoxazole, 6-methyl
PubChem CID 2724969
IUPAC Name 6-methyl-1,3-benzoxazole
SMILES CC1=CC2=C(C=C1)N=CO2

Specifications

CAS Min % 99.5
CAS Max % 100.0
Melting Point 29°C to 31°C
Color Amber
Infrared Spectrum Authentic
Assay Percent Range 99.5+%
Packaging Glass bottle
Quantity 10 g
Formula Weight 133.15
Percent Purity 99.5+%
Physical Form Low Melting Solid
Chemical Name or Material 6-Methylbenzoxazole
Show More Show Less
compliance-icons
Product Identifier
  • 6-Methylbenzoxazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.