missing translation for 'onlineSavingsMsg'
Learn More

6-Methylnicotinic acid, 99%

Product Code. p-3758443 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10227040 1 g Glass bottle 1g
10035963 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10227040 Supplier Thermo Scientific Acros Supplier No. 307620010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3222-47-7 | C7H7NO2 | 137.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3222-47-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.17
MDL Number MFCD00006341
InChI Key RZOKQIPOABEQAM-UHFFFAOYSA-N
Synonym 6-methylnicotinic acid, 3-pyridinecarboxylic acid, 6-methyl, 6-methyl nicotinic acid, 2-methylpyridine-5-carboxylic acid, 2-methyl-5-pyridinecarboxylic acid, 6-methyl-3-pyridinecarboxylic acid, 2-methyl-5-pyridine carboxylic acid, 6-methylnicotinate, 6-methylnicotinicacid, pubchem1218
PubChem CID 137860
IUPAC Name 6-methylpyridine-3-carboxylic acid
SMILES CC1=NC=C(C=C1)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 209°C to 213°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Formula Weight 137.17
Percent Purity 99%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 6-Methylnicotinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 6-Methylnicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 6-Methylnicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.