missing translation for 'onlineSavingsMsg'
Learn More

6-Methylpiperidin-2-one, TRC

Product Code. 30480425
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480425 100 mg 100mg
30474202 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30480425 Supplier Toronto Research Chemicals Supplier No. TRCM302710100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4775-98-8
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
Synonym 6-Methyl-2-piperidinone, 2-Pipecolin-6-one, Lactam-5-aminohexanoic Acid
IUPAC Name 6-methylpiperidin-2-one
SMILES CC1CCCC(=O)N1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H11NO/c1-5-3-2-4-6(8)7-5/h5H,2-4H2,1H3,(H,7,8)
Formula Weight 113.0841
Chemical Name or Material 6-Methylpiperidin-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.