missing translation for 'onlineSavingsMsg'
Learn More

6α-Methylprednisolone 21-hemisuccinate sodium salt, MedChemExpress

Product Code. 16458658
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458658 10 mg 10mg
16454537 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16458658 Supplier Medchem Express Supplier No. HYB1060
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 6α-Methylprednisolone 21-hemisuccinate sodium salt is a glucocorticoid of slightly longer half-life than that of Prednisolone.

6α-Methylprednisolone 21-hemisuccinate sodium salt is a glucocorticoid of slightly longer half-life than that of Prednisolone.

Specifications

Chemical Name or Material 6α-Methylprednisolone 21-hemisuccinate sodium salt
CAS 3-3-2375
Color White
Purity Grade Notes Research
Molecular Formula C26H33NaO8
Quantity 10 mg
Synonym Methylprednisolone sodium succinate 6-Methylprednisolone succinate sodium
Solubility Information H2O : 50 mg/mL (100.70 mM; Need ultrasonic) ∣DMSO : ≥ 25 mg/mL (50.35 mM)
SMILES C[C@@]12[C@](C(COC(CCC(O[Na])=O)=O)=O)(O)CC[C@@]1([H])[C@]3([H])C[C@H](C)C4=CC(C=C[C@]4(C)[C@@]3([H])[C@@H](O)C2)=O
Molecular Weight (g/mol) 496.53
Formula Weight 496.53
Percent Purity 95.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.