missing translation for 'onlineSavingsMsg'
Learn More

6-Methyluracil, TRC

Product Code. 30132859
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132859 50 g 50g
30005159 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132859 Supplier Toronto Research Chemicals Supplier No. TRCM33816050G
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-48-2
Molecular Formula C5 H6 N2 O2
Molecular Weight (g/mol) 126.11
Synonym 6-Methyl-2,4(1H,3H)-pyrimidinedione, Uracil, 6-methyl- (8CI), 6-Methyl-2,4(1H,3H)-pyrimidinedione, 2,4-Dihydroxy-6-methylpyrimidine, 6-Methylpyrimidine-2,4-diol, 6-Methyluracil, AWD 23-15, K 4, NSC 9456, Pseudothymine
IUPAC Name 6-methyl-1H-pyrimidine-2,4-dione
SMILES CC1=CC(=O)NC(=O)N1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C5H6N2O2/c1-3-2-4(8)7-5(9)6-3/h2H,1H3,(H2,6,7,8,9)
Formula Weight 126.0429
Percent Purity >95%
Chemical Name or Material 6-Methyluracil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.