missing translation for 'onlineSavingsMsg'
Learn More

6-Morpholin-4-yl-pyridine-3-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10553012
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10553012 250 mg Amber Glass Bottle 250mg
10438192 1 g Amber Glass Bottle 1g
10091404 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10553012 Supplier Thermo Scientific Maybridge Supplier No. CC17504CB
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 173282-60-5
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.22
MDL Number MFCD03659703
InChI Key VZEANTSDLFVWCK-UHFFFAOYSA-N
Synonym 6-morpholinonicotinaldehyde, 6-morpholin-4-yl-pyridine-3-carbaldehyde, 6-morpholin-4-ylnicotinaldehyde, 6-morpholin-4-yl pyridine-3-carbaldehyde, 6-morpholin-4-yl pyridine-3-carboxaldehyde, 6-morpholinylpyridine-3-carboxaldehyde, 6-morpholin-4-yl nicotinaldehyde, 6-morpholin-4-yl-pyridine-3-carbaldehyd, 6-4-morpholinyl-3-pyridinecarboxaldehyde, 3-pyridinecarboxaldehyde,6-4-morpholinyl
PubChem CID 2776463
SMILES O=CC1=CN=C(C=C1)N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 6-(morpholin-4-yl)pyridine-3-carbaldehyde
Formula Weight 192.22
Percent Purity 97%
Chemical Name or Material 6-Morpholinonicotinaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.