missing translation for 'onlineSavingsMsg'
Learn More

6-Morpholin-4-yl-pyridine-3-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10766455
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10766455 5 g Amber Glass Bottle 5g
10477612 250 mg Amber Glass Bottle 250mg
10171302 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10766455 Supplier Thermo Scientific Maybridge Supplier No. CC17503DE
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 337508-68-6
Molecular Formula C9H11ClN2O3S
Molecular Weight (g/mol) 262.708
MDL Number MFCD02681929
InChI Key FZQDEGBLWSULKG-UHFFFAOYSA-N
Synonym 6-morpholin-4-yl-pyridine-3-sulfonyl chloride, 6-morpholinopyridine-3-sulfonyl chloride, 6-morpholin-4-yl pyridine-3-sulfonyl chloride, 3-chlorosulphonyl-6-morpholin-4-yl pyridine, 3-pyridinesulfonyl chloride, 6-4-morpholinyl, 6-morpholin-4-yl pyridine-3-sulphonyl chloride, chloro 6-morpholin-4-yl 3-pyridyl sulfone, 6-4-morpholinyl-3-pyridinesulfonyl chloride, 3-pyridinesulfonylchloride, 6-4-morpholinyl, 4-5-chlorosulfonyl pyridin-2-yl morpholine
PubChem CID 2776462
IUPAC Name 6-morpholin-4-ylpyridine-3-sulfonyl chloride
SMILES C1COCCN1C2=NC=C(C=C2)S(=O)(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 262.72
Percent Purity 97%
Chemical Name or Material 6-Morpholin-4-yl-pyridine-3-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.