missing translation for 'onlineSavingsMsg'
Learn More

6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester, 95%

Product Code. 11327269
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11327269

Brand: Thermo Scientific Alfa Aesar H27430.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1032758-87-4 | C17H27BN2O4 | 334.223 g/mol

Used in the synthesis of organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of organic compounds.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1032758-87-4
Molecular Formula C17H27BN2O4
Molecular Weight (g/mol) 334.223
MDL Number MFCD09037484
InChI Key QGBJKHALDLKAGX-UHFFFAOYSA-N
Synonym tert-butyl methyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 6-boc-methylamino pyridine-3-boronic acid pinacol ester, 6-n-boc-methylamino pyridine-3-boronic acid pinacol ester, tert-butyl n-methyl-n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-methyl-n-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-methyl-n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 6-tert-butoxycarbonyl methyl amino pyridin-3-yl boronic acid pinacol ester, tert-butyl methyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-yl carbamate, tert-butylmethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate
PubChem CID 44541676
IUPAC Name tert-butyl N-methyl-N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)N(C)C(=O)OC(C)(C)C

Specifications

Melting Point 112°C to 119°C
Quantity 1 g
Formula Weight 334.22
Percent Purity 95%
Chemical Name or Material 6-(N-Boc-methylamino)pyridine-3-boronic acid pinacol ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.