missing translation for 'onlineSavingsMsg'
Learn More

BTX™ 6-needle Array Electrodes for BTX™ AgilePulse™ In Vivo

Product Code. 15407380
Change view
Click to view available options
Size (Metric) Gap:
4 mm
6 mm
Length (Metric):
2 mm
10 mm
16 mm
25 mm
Tip Type:
Fine Point
Trocar Point
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) Gap Length (Metric) Tip Type unitSize
15407380 4 mm 2 mm Fine Point Each
15417380 6 mm 2 mm Fine Point Each
15427380 6 mm 10 mm Fine Point Each
15447380 6 mm 16 mm Trocar Point Each
15437380 6 mm 25 mm Trocar Point Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15407380 Supplier BTX™ Supplier No. 470050
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

6-needle Array Electrodes for BTX™ AgilePulse™ In Vivo are miniature needles designed to provide highly-uniform electric fields in dermal or muscular tissue as part of the AgilePulse In Vivo Gene Delivery Electroporation System. Specially designed for gene and vaccine delivery.

The miniature parallel-needle array is inserted directly into the target site for fast, reliable in vivo electroporation. The electric fields produced are the closest approximation of parallel plate electrodes, treating approximately 80% of the target area with 95% of the applied electric field.

The needle array requires the needle array handle which includes the electrode connector cable. Several needle lengths and row spacing are available for various intra-dermal or intra-muscular applications. Needle Arrays can be used for up to 500 pulses with proper care and maintenance.

  • Uniform, reliable electric fields
  • Miniature needles minimize tissue trauma
  • Disposable and sterile
  • Medical-grade plastic and surgical steel construction
  • Safety-assured design
  • Multiple configurations, sizes to choose from

Tekniske data

For Use With (Application) Gene and Vaccine Delivery
For Use With (Equipment) AgilePulse™ in Vivo Gene Delivery Electroporation System
Material Stainless Steel
Product Line BTX Electroporation
Product Type Needle Array Electrodes
Description 6-needle Array, 4 mm gap, 2 mm length
Diameter (English) 0.01 in.
Diameter (Metric) 0.3 mm
Electrode Shape Straight/Needle array
Includes 4-needle Array
Length (English) 0.08 in.
Length (Metric) 2 mm
Quantity 1 Ea.
Size (Metric) Gap 4 mm
Tip Type Fine Point
Sterility Sterile
Connection Type Electrode Connector Cable
Pulse Form Square Wave
Vis mere Vis mindre

Intended for Research use only. Not for use in diagnostic or clinical procedures.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.