missing translation for 'onlineSavingsMsg'
Learn More

6-Nitrochromone, 97%

Product Code. 11456310
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456310 25 g 25g
11436310 1 g 1g
11446310 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456310 Supplier Thermo Scientific Alfa Aesar Supplier No. H27167.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51484-05-0 | C9H5NO4 | 191.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51484-05-0
Molecular Formula C9H5NO4
Molecular Weight (g/mol) 191.142
MDL Number MFCD02954226
InChI Key ORWADBVBOPTYQT-UHFFFAOYSA-N
Synonym 6-nitrochromone, 6-nitro-4h-chromen-4-one, 6-nitrobenzopyran-4-one, acmc-20alxw, 6-nitro-4-oxo-4h-1-benzopyran, 6-nitro-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,6-nitro
PubChem CID 847716
IUPAC Name 6-nitrochromen-4-one
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C=CO2

Specifications

Melting Point 172°C to 175°C
Quantity 25 g
Formula Weight 191.15
Percent Purity 97%
Chemical Name or Material 6-Nitrochromone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.