missing translation for 'onlineSavingsMsg'
Learn More

6-Nitropiperonyl alcohol, 98+%

Product Code. 11403657
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403657 5 g 5g
11413657 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11403657 Supplier Thermo Scientific Alfa Aesar Supplier No. A18140.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 15341-08-9 | C8H7NO5 | 197.15 g/mol

6-Nitropiperonyl alcohol is used as an organic chemical synthesis intermediate. It is also used as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Nitropiperonyl alcohol is used as an organic chemical synthesis intermediate. It is also used as a biochemical for proteomics research.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15341-08-9
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.15
MDL Number MFCD00005825
InChI Key LJXBRPHBDWIHPR-UHFFFAOYSA-N
Synonym 6-nitropiperonyl alcohol, 6-nitrobenzo d 1,3 dioxol-5-yl methanol, 3,4-methylenedioxy-6-nitrobenzyl alcohol, 6-nitro-3,4-methylenedioxybenzyl alcohol, 6-nitro-1,3-benzodioxol-5-yl methanol, 6-nitro-2h-1,3-benzodioxol-5-yl methanol, 6-nitro-1,3-benzodioxole-5-methanol, 1,3-benzodioxole-5-methanol,6-nitro, 1,3-benzodioxole-5-methanol, 6-nitro, 6-nitro-1,3-benzodioxol-5-yl methanol #
PubChem CID 519111
IUPAC Name (5-nitro-2H-1,3-benzodioxol-4-yl)methanol
SMILES OCC1=C2OCOC2=CC=C1[N+]([O-])=O

Specifications

Melting Point 121°C to 125°C
Quantity 5 g
Beilstein 13749
Sensitivity Light sensitive
Formula Weight 197.15
Percent Purity ≥98%
Chemical Name or Material 6-Nitropiperonyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.