missing translation for 'onlineSavingsMsg'
Learn More

6'-O-Apiosyl-5-O-Methylvisammioside, MedChemExpress

Product Code. 16452589
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452589 1 mg 1mg
16411708 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452589 Supplier Medchem Express Supplier No. HYN2295
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 6'-O-Apiosyl-5-O-Methylvisammioside is one of the components of the traditional Chinese medicine Kang-Jing.

6'-O-Apiosyl-5-O-Methylvisammioside is one of the components of the traditional Chinese medicine Kang-Jing.

Specifications

Chemical Name or Material 6'-O-Apiosyl-5-O-Methylvisammioside
CAS 139446-82-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H36O14
Quantity 1 mg
SMILES COC1=C(C[C@@H](C(C)(C)O[C@@H]([C@@H]([C@@H](O)[C@@H]2O)O)O[C@@H]2CO[C@H](OC[C@]3(O)CO)[C@@H]3O)O4)C4=CC(OC(C)=C5)=C1C5=O
Molecular Weight (g/mol) 584.57
Formula Weight 584.57
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.