missing translation for 'onlineSavingsMsg'
Learn More

6-O-Methylnorlaudanosoline Hydrochloride, TRC

Product Code. 30477616
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477616 10 mg 10mg
30474892 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477616 Supplier Toronto Research Chemicals Supplier No. TRCM32372510MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72072-58-3
Molecular Formula C17H20ClNO4
Molecular Weight (g/mol) 337.8
Synonym 4-[(1,2,3,4-Tetrahydro-7-hydroxy-6-methoxy-1-isoquinolinyl)methyl]-1,2-benzenediol Hydrochloride, (±)-4-[(1,2,3,4-Tetrahydro-7-hydroxy-6-methoxy-1-isoquinolinyl)methyl]-1,2-benzenediol Hydrochloride
IUPAC Name 4-[(7-hydroxy-6-methoxy-1,2,3,4-tetrahydroisoquinolin-1-yl)methyl]benzene-1,2-diol;hydrochloride
SMILES Cl.COc1cc2CCNC(Cc3ccc(O)c(O)c3)c2cc1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H19NO4.ClH/c1-22-17-8-11-4-5-18-13(12(11)9-16(17)21)6-10-2-3-14(19)15(20)7-10;/h2-3,7-9,13,18-21H,4-6H2,1H3;1H
Formula Weight 337.1081
Chemical Name or Material 6-O-Methylnorlaudanosoline Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.