missing translation for 'onlineSavingsMsg'
Learn More

6-Oxa-1-azaspiro[3.3]heptane hemioxalate, 95%

Product Code. p-7038647
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494098 250 mg 250mg
15404108 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494098 Supplier Thermo Scientific Alfa Aesar Supplier No. H57658.MD
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1380571-72-1 | C12H20N2O6 | 288.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1380571-72-1
Molecular Formula C12H20N2O6
Molecular Weight (g/mol) 288.3
MDL Number MFCD22373634
InChI Key UMKDEMSXCWMHRS-UHFFFAOYSA-N
Synonym 6-oxa-1-azaspiro 3.3 heptane oxalate 2:1, 6-oxa-1-azaspiro 3.3 heptane hemioxalate, 6-oxa-1-azaspiro 3.3 heptane oxalate 2, bis 6-oxa-1-azaspiro 3.3 heptane oxalate, 6-oxa-1-azaspiro 3.3 heptane,ethanedioate 2:1, oxalic acid-6-oxa-1-azaspiro 3.3 heptane 1/2, bis 6-oxa-1-azaspiro 3.3 heptane ; oxalic acid
PubChem CID 71299818
IUPAC Name 6-oxa-1-azaspiro[3.3]heptane;oxalic acid
SMILES C1CNC12COC2.C1CNC12COC2.C(=O)(C(=O)O)O

Specifications

Quantity 250 mg
Formula Weight 144.15
Percent Purity 95%
Chemical Name or Material 6-Oxa-1-azaspiro[3.3]heptane hemioxalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.