missing translation for 'onlineSavingsMsg'
Learn More

6-Oxo-1,6-dihydropyridazine-3-carboxylic acid monohydrate, 97%, Thermo Scientific™

Product Code. 10326394
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10326394

Brand: Thermo Scientific Maybridge BTB07063EB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 37972-69-3
Molecular Formula C5H3N2O3
Molecular Weight (g/mol) 139.09
MDL Number MFCD09064936
InChI Key GIFSROMQVPUQFK-UHFFFAOYSA-M
Synonym 6-hydroxypyridazine-3-carboxylic acid, 6-hydroxy-3-pyridazinecarboxylic acid, 6-oxo-1,6-dihydropyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-pyridazine-3-carboxylic acid, 3-hydroxypyridazine-6-carboxylic acid, 3-pyridazinecarboxylic acid, 1,6-dihydro-6-oxo, 6-hydroxy-pyridazine-3-carboxylic acid, 6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid, 3-pyridazinecarboxylicacid, 1,6-dihydro-6-oxo, 6-oxohydropyridazine-3-carboxylic acid
PubChem CID 305970
IUPAC Name 6-oxo-1H-pyridazine-3-carboxylic acid
SMILES [O-]C(=O)C1=NNC(=O)C=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 158.12
Percent Purity 97%
Chemical Name or Material 6-Hydroxy-3-pyridazinecarboxylic acid monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.