missing translation for 'onlineSavingsMsg'
Learn More

6-Phenoxynicotinic acid, 97%, Thermo Scientific™

Product Code. 10722093
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10722093 250 mg 250mg
10059221 1 g 1g
10457744 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10722093 Supplier Thermo Scientific Maybridge Supplier No. CC19601CB
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 51362-38-0
Molecular Formula C12H9NO3
Molecular Weight (g/mol) 215.208
MDL Number MFCD02682068
InChI Key GFEUNYLQJDQNAN-UHFFFAOYSA-N
Synonym 6-phenoxynicotinic acid, 3-pyridinecarboxylic acid, 6-phenoxy, 6-phenoxynicotinicacid, 6-phenoxy-nicotinic acid, 2-phenoxypyridine-5-carboxylic acid, 3-pyridinecarboxylicacid, 6-phenoxy, 6-phenyloxy-3-pyridinecarboxylic acid
PubChem CID 2776497
IUPAC Name 6-phenoxypyridine-3-carboxylic acid
SMILES C1=CC=C(C=C1)OC2=NC=C(C=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 215.21
Percent Purity 97%
Chemical Name or Material 6-Phenoxynicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.