missing translation for 'onlineSavingsMsg'
Learn More

6-Phenoxypyridin-3-amine, TRC

Product Code. 30423772
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423772 500 mg 500mg
30410074 1 g 1g
30412683 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423772 Supplier Toronto Research Chemicals Supplier No. TRCP318898500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25194-67-6
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.2
Synonym 6-Phenoxy-3-pyridinamine, 2-Phenoxy-5-aminopyridine, 3-Amino-6-phenoxypyridine, 5-Amino-2-phenoxypyridine
IUPAC Name 6-phenoxypyridin-3-amine
SMILES Nc1ccc(Oc2ccccc2)nc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H10N2O/c12-9-6-7-11(13-8-9)14-10-4-2-1-3-5-10/h1-8H,12H2
Formula Weight 186.0793
Chemical Name or Material 6-Phenoxypyridin-3-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.