missing translation for 'onlineSavingsMsg'
Learn More

6-Phenyl-1,3,5-triazine-2,4-diamine, TRC

Product Code. 30180151
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180151 250 mg 250mg
30180143 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180151 Supplier Toronto Research Chemicals Supplier No. TRCP321073250MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-76-9
Molecular Formula C9 H9 N5
Molecular Weight (g/mol) 187.2
Synonym s-Triazine, 2,4-diamino-6-phenyl- (6CI,8CI), 6-Phenyl-1,3,5-triazine-2,4-diamine, 2,4-Diamino-6-phenyl-1,3,5-triazine, 2,4-Diamino-6-phenyl-s-triazine, 2,6-Diamino-4-phenyl-1,3,5-triazine, 2-Phenyl-4,6-diamino-1,3,5-triazine, 2-Phenyl-4,6-diamino-s-triazine, 4,6-Diamino-2-phenyl-s-triazine, 6-Phenyl-2,4-diamino-1,3,5-triazine, Benzoguanamine, Benzoguanimine, Benzoquanamine, ENT 60118, NSC 3267, Nikaganamine
IUPAC Name 6-phenyl-1,3,5-triazine-2,4-diamine
SMILES Nc1nc(N)nc(n1)c2ccccc2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H9N5/c10-8-12-7(13-9(11)14-8)6-4-2-1-3-5-6/h1-5H,(H4,10,11,12,13,14)
Formula Weight 187.0858
Chemical Name or Material Benzoguanamine (6-Phenyl-1,3,5-triazine-2,4-diamine)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.