missing translation for 'onlineSavingsMsg'
Learn More

6-Phenylnicotinic acid, 95%, Thermo Scientific™

Product Code. 10712473
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10712473 250 mg Amber Glass Bottle 250mg
10487222 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10712473 Supplier Thermo Scientific Maybridge Supplier No. CC21101CB
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 29051-44-3
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD03086128
InChI Key DLFLQXUYRFIFOK-UHFFFAOYSA-N
Synonym 6-phenylnicotinic acid, 3-pyridinecarboxylic acid, 6-phenyl, 6-phenyl-nicotinic acid, 2-phenyl pyridine-5-carboxylic acid, 6-phenyl nicotinic acid, acmc-1cmxf, 2-phenyl-5-carboxypyridine, 3-carboxy-6-phenylpyridine, 2-phenyl-5-pyridinecarboxylic acid, 6-phenyl-3-pyridinecarboxylic acid
PubChem CID 120118
IUPAC Name 6-phenylpyridine-3-carboxylic acid
SMILES C1=CC=C(C=C1)C2=NC=C(C=C2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.21
Percent Purity 95%
Chemical Name or Material 6-Phenylnicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.