missing translation for 'onlineSavingsMsg'
Learn More

6-Phenylnicotinonitrile, 97%, Thermo Scientific™

Product Code. 10547854
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10547854 250 mg 250mg
10295404 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10547854 Supplier Thermo Scientific Maybridge Supplier No. CC21116CB
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 39065-54-8
Molecular Formula C12H8N2
Molecular Weight (g/mol) 180.21
MDL Number MFCD06637420
InChI Key OSRYUVXCTNUGRM-UHFFFAOYSA-N
PubChem CID 12576518
IUPAC Name 6-phenylpyridine-3-carbonitrile
SMILES C1=CC=C(C=C1)C2=NC=C(C=C2)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 6-phenylnicotinonitrile, 5-cyano-2-phenylpyridine, 3-pyridinecarbonitrile,6-phenyl, 3-pyridinecarbonitrile, 6-phenyl, 6-phenylniocotinonitrile, 2-phenyl-5-cyanopyridine, 3-cyano-6-phenylpyridine, 6-phenyl-3-pyridinecarbonitrile
Formula Weight 180.21
Percent Purity 97%
Chemical Name or Material 6-Phenylnicotinonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.