missing translation for 'onlineSavingsMsg'
Learn More

6-piperidin-1-ylnicotinaldehyde, Thermo Scientific™

Product Code. 10159212
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10159212 1 g 1g
10002454 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10159212 Supplier Thermo Scientific Maybridge Supplier No. MO07199DA
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 241816-11-5
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.246
InChI Key RKIMIISNVCLRLA-UHFFFAOYSA-N
Synonym 6-piperidin-1-ylnicotinaldehyde, 6-piperidin-1-yl nicotinaldehyde, 6-piperidin-1-yl pyridine-3-carbaldehyde, 6-piperidinonicotinaldehyde, 6-1-piperidinyl nicotinaldehyde, 6-1-piperidinyl pyridine-3-carboxaldehyde, 6-piperidylpyridine-3-carbaldehyde, acmc-20alxp, 3-pyridinecarboxaldehyde,6-1-piperidinyl, 3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-5'-carbaldehyde
PubChem CID 2763980
IUPAC Name 6-piperidin-1-ylpyridine-3-carbaldehyde
SMILES C1CCN(CC1)C2=NC=C(C=C2)C=O

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 190.24
Percent Purity 97%
Chemical Name or Material 6-Piperidin-1-ylnicotinaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.