missing translation for 'onlineSavingsMsg'
Learn More

(6-Piperidinopyrid-2-yl)methanol, 97%, Thermo Scientific™

Product Code. 10680454
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10680454 1 g Amber Glass Bottle 1g
10245274 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10680454 Supplier Thermo Scientific Maybridge Supplier No. CC38109DA
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 869901-07-5
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
MDL Number MFCD08690242
InChI Key WLHPFWHASVOLSO-UHFFFAOYSA-N
Synonym 6-piperidinopyrid-2-yl methanol, 6-piperidin-1-yl pyridin-2-yl methanol, 6-piperidyl-2-pyridyl methan-1-ol, 2-pyridinemethanol,6-1-piperidinyl, 6-piperidin-1-ylpyridin-2-yl methanol, 2-hydroxymethyl-6-piperidin-1-ylpyridine
PubChem CID 18525733
IUPAC Name (6-piperidin-1-ylpyridin-2-yl)methanol
SMILES C1CCN(CC1)C2=CC=CC(=N2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 192.26
Percent Purity 97%
Chemical Name or Material (6-Piperidinopyrid-2-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.