missing translation for 'onlineSavingsMsg'
Learn More

(6-Piperidinopyrid-2-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10398313
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10398313 250 mg 250mg
10149162 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10398313 Supplier Thermo Scientific Maybridge Supplier No. CC38113CB
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 868755-51-5
Molecular Formula C11H17N3
Molecular Weight (g/mol) 191.28
MDL Number MFCD08690243
InChI Key AMSLKMQNMJEDRH-UHFFFAOYSA-N
Synonym 6-piperidinopyrid-2-yl methylamine, 6-piperidin-1-yl pyridin-2-yl methanamine, 1-6-piperidin-1-yl pyridin-2-yl methanamine, 6-piperidyl-2-pyridyl methylamine, 2-pyridinemethanamine,6-1-piperidinyl, 6-piperidin-1-yl pyridin-2-yl methylamine, 1-6-piperidin-1-yl pyridin-2-yl methylamine
PubChem CID 18525734
IUPAC Name (6-piperidin-1-ylpyridin-2-yl)methanamine
SMILES NCC1=NC(=CC=C1)N1CCCCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 191.28
Percent Purity 97%
Chemical Name or Material (6-Piperidinopyrid-2-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.