missing translation for 'onlineSavingsMsg'
Learn More

(6-Pyrrolidin-1-ylpyrid-2-yl)methanol, 97%, Thermo Scientific™

Product Code. 10587084
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10587084 250 mg Amber Glass Bottle 250mg
10285744 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10587084 Supplier Thermo Scientific Maybridge Supplier No. CC38009CB
 Restricted Item
Estimated Shipment: 15-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 868755-48-0
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD09702357
InChI Key INCLVFBQYYTJLC-UHFFFAOYSA-N
Synonym 6-pyrrolidin-1-ylpyrid-2-yl methanol, 6-pyrrolidin-1-yl pyridin-2-yl methanol, 2-pyridinemethanol,6-1-pyrrolidinyl, 6-pyrrolidin-1-ylpyridin-2-yl methanol, 6-pyrrolidinyl-2-pyridyl methan-1-ol, 1-2-hydroxymethyl pyridin-2-yl pyrrolidine, 2-hydroxymethyl-6-pyrrolidin-1-yl pyridine
PubChem CID 24229483
IUPAC Name (6-pyrrolidin-1-ylpyridin-2-yl)methanol
SMILES C1CCN(C1)C2=CC=CC(=N2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material (6-Pyrrolidin-1-ylpyrid-2-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.