missing translation for 'onlineSavingsMsg'
Learn More

6-Pyrrolidin-1-ylpyridine-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10675155
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10675155 1 g Amber Glass Bottle 1g
10245504 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10675155 Supplier Thermo Scientific Maybridge Supplier No. CC38001DA
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 450368-20-4
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.22
MDL Number MFCD09064946
InChI Key HXPHLYQBFPDHNK-UHFFFAOYSA-N
PubChem CID 23509740
SMILES OC(=O)C1=NC(=CC=C1)N1CCCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 6-1-pyrrolidyl pyridine-2-carboxylic acid, 6-pyrrolidin-1-yl picolinic acid, 6-pyrrolidin-1-yl pyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 6-1-pyrrolidinyl, 6-pyrrolidinylpyridine-2-carboxylic acid, 6-1-pyrrolidyl pyridine-2-carboxylicacid, 2-pyridinecarboxylicacid,6-1-pyrrolidinyl, 6-1-pyrrolidinyl-2-pyridinecarboxylic acid
IUPAC Name 6-pyrrolidin-1-ylpyridine-2-carboxylic acid
Formula Weight 192.22
Percent Purity 97%
Chemical Name or Material 6-Pyrrolidin-1-ylpyridine-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.