missing translation for 'onlineSavingsMsg'
Learn More

6-Quinoxalinamine, TRC

Product Code. 17962815
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962815 1 g 1g
17920345 5 g 5g
17914775 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17962815 Supplier Toronto Research Chemicals Supplier No. TRCQ7500101G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6298-37-9
Molecular Formula C8 H7 N3
Molecular Weight (g/mol) 145.16
Synonym 6-Quinoxalinamine, Quinoxaline, 6-amino- (6CI,7CI,8CI), (Quinoxalin-6-yl)amine, 6-Aminoquinoxaline, 6-Quinoxalylamine, NSC 41810
IUPAC Name quinoxalin-6-amine
SMILES Nc1ccc2nccnc2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H7N3/c9-6-1-2-7-8(5-6)11-4-3-10-7/h1-5H,9H2
Formula Weight 145.064
Chemical Name or Material Quinoxalin-6-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.