missing translation for 'onlineSavingsMsg'
Learn More

6(R)-[2-(8(S)-Hydroxy]-2(S),6(R)-dimethyl-1,2,6,7,8,8a(R)-hexahydro-1(S)-naphthyl]ethyl-4(R)-hydroxy-3,4,5,6-tetrahydro-2H-pyran-2-one, TRC

Product Code. 30408292
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30408292 10 mg
30395908 25 mg
30399094 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408292 Supplier Toronto Research Chemicals Supplier No. TRCH94162510MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79952-42-4
Molecular Formula C19 H28 O4
Molecular Weight (g/mol) 320.42
Synonym 2H-Pyran-2-one, 6-[2-[(1S,2S,6R,8S,8aR)-1,2,6,7,8,8a-hexahydro-8-hydroxy-2,6-dimethyl-1-naphthalenyl]ethyl]tetrahydro-4-hydroxy-, (4R,6R)-, 2H-Pyran-2-one, 6-[2-(1,2,6,7,8,8a-hexahydro-8-hydroxy-2,6-dimethyl-1-naphthalenyl)ethyl]tetrahydro-4-hydroxy-, [1S-[1α(4S*,6S*),2α,6β,8β,8aα]]-, (4R,6R)-6-[2-[(1S,2S,6R,8S,8aR)-1,2,6,7,8,8a-Hexahydro-8-hydroxy-2,6-dimethyl-1-naphthalenyl]ethyl]tetrahydro-4-hydroxy-2H-pyran-2-one, Antibiotic MB 530A, Lovastatin diol lactone, Monacolin J
IUPAC Name (4R,6R)-6-[2-[(1S,2S,6R,8S,8aR)-8-hydroxy-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]ethyl]-4-hydroxyoxan-2-one
SMILES C[C@@H]1C[C@H](O)[C@@H]2[C@@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H](C)C=CC2=C1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H28O4/c1-11-7-13-4-3-12(2)16(19(13)17(21)8-11)6-5-15-9-14(20)10-18(22)23-15/h3-4,7,11-12,14-17,19-21H,5-6,8-10H2,1-2H3/t11-,12-,14+,15+,16-,17-,19-/m0/s1
Formula Weight 320.1988
Chemical Name or Material Lovastatin diol lactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.