missing translation for 'onlineSavingsMsg'
Learn More

6-(Tetrahydropyran-4-yloxy)nicotinonitrile, 97%, Thermo Scientific™

Product Code. 10255564
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10255564 5 g Amber Glass Bottle 5g
10092304 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10255564 Supplier Thermo Scientific Maybridge Supplier No. CC58316DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 884507-60-2
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD09702388
InChI Key CCEWHGWDICBASG-UHFFFAOYSA-N
Synonym 6-tetrahydropyran-4-yloxy nicotinonitrile, 6-tetrahydro-2h-pyran-4-yloxy pyridine-3-carbonitrile, 3-pyridinecarbonitrile,6-tetrahydro-2h-pyran-4-yl oxy, 6-oxan-4-yloxy pyridine-3-carbonitrile, 6-oxan-4-yl oxy pyridine-3-carbonitrile, 6-tetrahydro-2h-pyran-4-yl oxy nicotinonitrile, 6-2h-3,4,5,6-tetrahydropyran-4-yloxy pyridine-3-carbonitrile
PubChem CID 24229580
IUPAC Name 6-(oxan-4-yloxy)pyridine-3-carbonitrile
SMILES N#CC1=CN=C(OC2CCOCC2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 204.23
Percent Purity 97%
Chemical Name or Material 6-(Tetrahydropyran-4-yloxy)nicotinonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.