missing translation for 'onlineSavingsMsg'
Learn More

(6-Thien-2-ylpyrid-3-yl)methanol, 95%, Thermo Scientific™

Product Code. 10347104
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Unit Size:
250mg
Product Code. Quantity Packaging unitSize
10347104 250 mg Amber Glass Bottle 250mg
1 options
This item is not returnable. View return policy

Product Code. 10347104

Brand: Thermo Scientific Maybridge CC54809CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 198078-57-8
Molecular Formula C10H9NOS
Molecular Weight (g/mol) 191.248
MDL Number MFCD06410427
InChI Key OLHQRPWNQRBQMA-UHFFFAOYSA-N
Synonym 6-thien-2-ylpyrid-3-yl methanol, 6-thiophen-2-yl pyridin-3-yl methanol, 3-pyridinemethanol,6-2-thienyl, 2-thien-2-yl-5-hydroxymethylpyridine, 6-2-thienyl-3-pyridyl methan-1-ol
PubChem CID 15411145
IUPAC Name (6-thiophen-2-ylpyridin-3-yl)methanol
SMILES C1=CSC(=C1)C2=NC=C(C=C2)CO

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 191.25
Percent Purity 95%
Chemical Name or Material (6-Thien-2-ylpyrid-3-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.