missing translation for 'onlineSavingsMsg'
Learn More

6-(Trifluoromethyl)indole, 97%

Product Code. 11383896
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11383896 250 mg 250mg
11373896 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383896 Supplier Thermo Scientific Alfa Aesar Supplier No. L17009.MD
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13544-43-9 | C9H6F3N | 185.149 g/mol

6-(Trifluoromethyl)indole is used in pharmaceutical intermediates, liquid crystal intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-(Trifluoromethyl)indole is used in pharmaceutical intermediates, liquid crystal intermediates

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13544-43-9
Molecular Formula C9H6F3N
Molecular Weight (g/mol) 185.149
MDL Number MFCD00272316
InChI Key BPYBYPREOVLFED-UHFFFAOYSA-N
Synonym 6-trifluoromethyl indole, 6-trifluoromethylindole, 6-trifluoromethyl-1h-indole, 1h-indole, 6-trifluoromethyl, pubchem1713, acmc-1bscm, intermediates-zcf02044, 6-trifluoromethyl-indole, ksc910g4n
PubChem CID 2777523
IUPAC Name 6-(trifluoromethyl)-1H-indole
SMILES C1=CC(=CC2=C1C=CN2)C(F)(F)F

Specifications

Melting Point 101°C to 105°C
Quantity 250 mg
Beilstein 1242785
Solubility Information Insoluble in water.
Formula Weight 185.15
Percent Purity 97%
Chemical Name or Material 6-(Trifluoromethyl)indole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.