Learn More
Ohaus™ 6 x Centrifuge Tube Adapters, Open Bottom, Frontier™ Fixed Angle Rotors
Product Code. 15856632 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
0.2/0.4 mL
0.5 mL
1.5/2.0 mL
Unit Size:
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
15856632 1.5/2.0 mL Pack of 6
15836632 0.2/0.4 mL Pack of 6
15846632 0.5 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15856632 Supplier Ohaus™ Supplier No. 30130886
  • ONLINE EXCLUSIVE PRICE
    101.00 €
    74.68 € / Pack of 6
    Save 26.32 €
    26% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Facilitate centrifugation of tubes smaller than the original, provided carrier space for an array of Frontier™ fixed angle rotors.
  • Accommodates centrifuge tubes that are smaller than the space provided in the rotor carrier
  • Flat-, open-bottom carrier.
  • Suitable for use in the following Frontier™ rotors:
    • R-A30x2/13MS
    • R-A24x2/15MS
    • R-A24x2/15M
    • R-A12x2/18MI
    • R-A24x2/16MI
    • R-A30x2/14MIS
    • R-A44x2/16MI
    • R-A24x2/16MIS
TRUSTED_SUSTAINABILITY
Specifications
Capacity 1.5/2.0 mL
Max. Tube Diameter 11 mm
For Use With (Equipment) Frontier Fixed-Angle Rotors
Description Adapter, D11mm, 1.5/2.0ml Tubes x6
No. of Places 1
No. per Pack 6
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.