missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 680C91

Description
680C91 is a potent and selective inhibitor of tryptophan 2,3-dioxygenase (TDO) (Ki = 51 nM). Exhibits no activity against indoleamine 2,3-dioxygenase, monoamine oxidase A and B, 5-HT uptake or 5-HT1A, 1D, 2A and 2C receptors. Produces large increases in brain tryptophan and serotonin in vitro and in vivo in the rat.
Specifications
Specifications
| Chemical Name or Material | 6-Fluoro-3-[(1E)-2-(3-pyridinyl)ethenyl]-1H-indole |
| CAS | 163239-22-3 |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Molecular Formula | C15H11FN2 |
| Formula Weight | Observed MW: 238.26 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?