missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 680C91
GREENER_CHOICE

Product Code. 18757596
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18757596 5 mg 5mg
18727826 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18757596 Supplier Tocris Bioscience™ Supplier No. 4392/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective tryptophan 2,3-dioxygenase (TDO) inhibitor

680C91 is a potent and selective inhibitor of tryptophan 2,3-dioxygenase (TDO) (Ki = 51 nM). Exhibits no activity against indoleamine 2,3-dioxygenase, monoamine oxidase A and B, 5-HT uptake or 5-HT1A, 1D, 2A and 2C receptors. Produces large increases in brain tryptophan and serotonin in vitro and in vivo in the rat.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Fluoro-3-[(1E)-2-(3-pyridinyl)ethenyl]-1H-indole
CAS 163239-22-3
Assay Percent Range >98%
Quantity 5 mg
Molecular Formula C15H11FN2
Formula Weight Observed MW: 238.26
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.