missing translation for 'onlineSavingsMsg'
Learn More

(6Alpha,9Beta,11Beta,16Alpha)-9,11-Epoxy-6-fluoro-21-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, TRC

Product Code. 30012339
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012339 25 mg 25mg
30003875 2.5 mg 2.5mg
30006700 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012339 Supplier Toronto Research Chemicals Supplier No. TRCE58928525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68352-03-4
Molecular Formula C24 H29 F O6
Molecular Weight (g/mol) 432.48
Synonym Pregna-1,4-diene-3,20-dione, 9,11-epoxy-6-fluoro-21-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-, (6α,9β,11β,16α)-, (6α,9β,11β,16α)-9,11-Epoxy-6-fluoro-21-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, 9,11β-Epoxy-6α-fluoro-21-hydroxy-16α,17-(1-methylethylidenedioxy)-9β-pregna-1,4-diene-3,20-dione
SMILES CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]46O[C@H]6C[C@]3(C)[C@@]2(O1)C(=O)CO

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C24H29FO6/c1-20(2)29-18-9-13-14-8-16(25)15-7-12(27)5-6-21(15,3)23(14)19(30-23)10-22(13,4)24(18,31-20)17(28)11-26/h5-7,13-14,16,18-19,26H,8-11H2,1-4H3/t13-,14-,16-,18+,19-,21-,22-,23+,24+/m0/s1
Formula Weight 432.1948
Chemical Name or Material 9,11beta-Epoxy-6alpha-fluoro-21-hydroxy-16alpha,17-(1-methylethylidenedioxy)-9beta-pregna-1,4-diene-3,20-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.