missing translation for 'onlineSavingsMsg'
Learn More

6alpha-Methyl Prednisolone Aceponate, TRC

Product Code. 30008250
Change view
Click to view available options
Quantity:
25 mg
250 mg
1 g
Unit Size:
1g
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008250 25 mg 25mg
30008210 250 mg 250mg
30004555 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008250 Supplier Toronto Research Chemicals Supplier No. TRCM32572525MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86401-95-8
Molecular Formula C27 H36 O7
Molecular Weight (g/mol) 472.57
Synonym Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-11-hydroxy-6-methyl-17-(1-oxopropoxy)-, (6α,11β)-, (6α,11β)-21-(Acetyloxy)-11-hydroxy-6-methyl-17-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, Advantan, Methylprednisolone aceponate, ZK 91588, 6α-Methylprednisolone aceponate
IUPAC Name [(6S,8S,9S,10R,11S,13S,14S,17R)-17-(2-acetyloxyacetyl)-11-hydroxy-6,10,13-trimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl] propanoate
SMILES CCC(=O)O[C@@]1(CC[C@H]2[C@@H]3C[C@H](C)C4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)COC(=O)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C27H36O7/c1-6-23(32)34-27(22(31)14-33-16(3)28)10-8-19-18-11-15(2)20-12-17(29)7-9-25(20,4)24(18)21(30)13-26(19,27)5/h7,9,12,15,18-19,21,24,30H,6,8,10-11,13-14H2,1-5H3/t15-,18-,19-,21-,24+,25-,26-,27-/m0/s1
Formula Weight 472.2461
Percent Purity >95%
Chemical Name or Material 6-alpha-Methyl Prednisolone Aceponate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.