missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 6Ckine (CCL21) Mouse ProcartaPlex™ Simplex Kit

Product Code. 17882984
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17882984 96 Tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17882984 Supplier Invitrogen™ Supplier No. EPX01026101901
  • ONLINE EXCLUSIVE PRICE
    349.00 €
    335.04 € / Pack of 96
    Save 13.96 €
    4% Off
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Mouse 6Ckine (CCL21) ProcartaPlex Simplex Kit measures 6Ckine (CCL21) protein and is designed to be combinable with other Simplex kits so that users can create their own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation.

The Mouse 6Ckine (CCL21) ProcartaPlex Simplex Kit measures 6Ckine (CCL21) protein and is designed to be combinable with other Simplex kits so that users can create their own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

About ProcartaPlex assays for the Luminex platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

Features include:
• More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
• Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 65 protein targets in a single 25–50 μL sample—from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine).

TRUSTED_SUSTAINABILITY

Specifications

Product Type Simplex Kit
Quantity 96 Tests
Species Mouse
Combinability Combinable
Bead Type Magnetic
Assay Sensitivity As determined for Lot 1: 0.012
Assay Range As determined for Lot 1: 0.012–50 pg/mL
Format Simplex Kit
For Use With (Equipment) Luminex™ Instruments
High-throughput Compatibility Yes (384-well Mix and Match panel)
Interassay CV As determined for Lot 1: 6.9%
Intraassay CV As determined for Lot 1: 3%
Product Line ProcartaPlex
Sample Type Serum, Plasma, Cell Culture Supernatants (CCS)
Sample Volume Serum, Plasma: 25 μL
CCS: 50 μL
Content And Storage • 1 vial Capture Beads (50X)
• 1 vial Biotinylated Detection Antibody (50X)
• 2 vials Standard Mix (lyophilized)

Store at 2–8°C.

Shipping Condition Wet Ice
Research Area Chemokines, Immunology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.