Learn More
Univet™ 6X3 Goggles for PPE
Product Code. 17960862
Change view
Click to view available options
UV Protection:
UV380
Unit Size:
Pack of 5
Product Code. UV Protection Unit Size
17960862 UV380 Pack of 5
1 options
Product Code. 17960862 Supplier Univet™ Supplier No. 6X3.00.00.00
  • ONLINE EXCLUSIVE PRICE
    94.31 €
    79.65 € / Pack of 5
    Save 14.66 €
    15% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
These 6X3 Goggles offer all-round protection and high performance, even when worn with other PPE. They can be worn comfortably over prescription glasses and still provide optimum comfort.
  • Certified according to EN166 standard, point 3 and 4 for protection against droplets, splashes and large dust particles
  • Panoramic lens offers ideal peripheral vision and is easily integrated with prescription glasses
  • Fast interchange lens system
  • Frame engineered for a soft facial support
  • Compatible with filtering half masks
  • Lens with anti-fog and anti-scratch ultra performing Vanguard UDC treatment
Specifications
Shape Rectangular
Lens Coating Anti-fog, Scratch-resistant
Lens Tint None
Product Type Heat-resistant Safety Goggles
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material PCB
UV Protection UV380
Frame Type Frameless
Autoclavable Not Autoclavable