missing translation for 'onlineSavingsMsg'
Learn More

7-(2,3-Dihydroxypropyl)theophylline, 99%

Product Code. 10080830 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10080830

Brand: Thermo Scientific Acros 115051000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 479-18-5 | C10H14N4O4 | 254.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479-18-5
Molecular Formula C10H14N4O4
Molecular Weight (g/mol) 254.24
MDL Number MFCD00005759
InChI Key KSCFJBIXMNOVSH-UHFFFAOYSA-N
Synonym dyphylline, diprophylline, diphylline, 7-2,3-dihydroxypropyl theophylline, diprophyllin, glyphylline, neothylline, lufyllin, aristophyllin, diprofilline
PubChem CID 3182
ChEBI CHEBI:4728
IUPAC Name 7-(2,3-dihydroxypropyl)-1,3-dimethylpurine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CC(CO)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 160°C to 165°C
Color White
Loss on Drying 0.5% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 3526
Solubility Information Solubility in water: 1g/3mL (25 c). Other solubilities: 2g/100mL ethanol,1g/100mL chloroform
Formula Weight 254.24
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 7-(2, 3-Dihydroxypropyl)theophylline, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 7-(2,3-Dihydroxypropyl)theophylline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.