missing translation for 'onlineSavingsMsg'
Learn More

7-(2,3-Dihydroxypropyl)theophylline, 99%

Product Code. 10626112 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10626112 500 g Glass bottle 500g
10080830 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10626112 Supplier Thermo Scientific Acros Supplier No. 115055000
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 479-18-5 | C10H14N4O4 | 254.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479-18-5
Molecular Formula C10H14N4O4
Molecular Weight (g/mol) 254.24
MDL Number MFCD00005759
InChI Key KSCFJBIXMNOVSH-UHFFFAOYSA-N
Synonym dyphylline, diprophylline, diphylline, 7-2,3-dihydroxypropyl theophylline, diprophyllin, glyphylline, neothylline, lufyllin, aristophyllin, diprofilline
PubChem CID 3182
ChEBI CHEBI:4728
IUPAC Name 7-(2,3-dihydroxypropyl)-1,3-dimethylpurine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CC(CO)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 160°C to 165°C
Color White
Loss on Drying 0.5% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 500 g
Merck Index 15, 3526
Solubility Information Solubility in water: 1g/3mL (25 c). Other solubilities: 2g/100mL ethanol,1g/100mL chloroform
Formula Weight 254.24
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 7-(2, 3-Dihydroxypropyl)theophylline, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 7-(2,3-Dihydroxypropyl)theophylline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.