missing translation for 'onlineSavingsMsg'
Learn More

7-(2-Chloroethyl)theophylline, 97%, Thermo Scientific™

Product Code. 10051800 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10051800 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10051800 Supplier Thermo Scientific Acros Supplier No. 155750050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 5878-61-5
CAS Min % 96.0
CAS Max % 100.0
Melting Point 122°C to 127°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C9H11ClN4O2
MDL Number MFCD00005760
Quantity 5 g
Merck Index 10, 2109
Synonym 7-2-chloroethyl theophylline, benaphyllin, eupnophile, 7-chloroethyl theophylline, theophylline, 7-2-chloroethyl, beta-chloroethyltheophylline, unii-y7202un6b7, 7-beta-chlorethyl theophylline, 7-chloroethyltheophylline, 1h-purine-2,6-dione, 7-2-chloroethyl-3,7-dihydro-1,3-dimethyl
InChI Key QCIARNIKNKKHFH-UHFFFAOYSA-N
SMILES CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CCCl
IUPAC Name 7-(2-chloroethyl)-1,3-dimethylpurine-2,6-dione
Molecular Weight (g/mol) 242.67
PubChem CID 1882
Formula Weight 242.67
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 7-(2-Chloroethyl)theophylline, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 7-(2-Chloroethyl)theophylline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.