missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 7,8-Dihydroxyflavone
GREENER_CHOICE

Product Code. 18003852
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18003852 10 mg 10mg
18076002 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18003852 Supplier Tocris Bioscience™ Supplier No. 3826/10
  • ONLINE EXCLUSIVE PRICE
    240.00 €
    226.80 € / 10mg
    Save 13.20 €
    5% Off
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

TrkB agonist

Tyrosine kinase receptor B (TrkB) agonist that binds to the extracellular domain of the receptor (Kd = 320 nM). Inhibits glutamate-triggered apoptosis in hippocampal neurons in vitro and in vivo. Exhibits neuroprotective effects in an HD mouse model.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38183-03-8
Molecular Formula C15H10O4

Specifications

Quantity 10 mg
Formula Weight 254.24
Purity >98%
Molecular Weight (g/mol) 254.24
Chemical Name or Material 7, 8-Dihydroxyflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.