missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 7-Amino-4-methylcoumarin

Product Code. 16464657
Encompass_Preferred
Change view
Click to view available options
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16464657 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16464657 Supplier MedChemExpress Supplier No. HYD0027

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress 7-amino-4-methylcoumarin (Coumarin 120) has been used as a fluorogenic probe for many serine proteases.

Specifications

Quantity 50 mg
Product Type Dye Reagents
Molecular Weight (g/mol) 175.18 g/mol
Solubility DMSO : 100 mg/mL (570.84 mM; Need ultrasonic)
Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
Target HBV
CAS 26093-31-2
Synonym Coumarin 120 ∣ AMC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.