missing translation for 'onlineSavingsMsg'
Learn More

7-Aminocephalosporanic acid, 95-102%

Product Code. 10513361 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10513361 1 g Glass bottle 1g
10634692 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10513361 Supplier Thermo Scientific Acros Supplier No. 184410010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 957-68-6 | C10H12N2O5S | 272.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 957-68-6
Molecular Formula C10H12N2O5S
Molecular Weight (g/mol) 272.27
MDL Number MFCD00005177
InChI Key HSHGZXNAXBPPDL-HZGVNTEJSA-N
Synonym 7-aminocephalosporanic acid, 7-aca, 7-aminocephalosporinic acid, 7beta-aminocephalosporanic acid, 7-acs, unii-9xi67897rg, 7r-7-aminocephalosporanic acid, 7r-7-aminocephalosporanate, cephalosporanic acid, 7-amino, 6r,7r-3-acetoxymethyl-7-amino-8-oxo-5-thia-1-azabicyclo 4.2.0 oct-2-ene-2-carboxylic acid
PubChem CID 441328
ChEBI CHEBI:2255
IUPAC Name (6R,7R)-3-(acetyloxymethyl)-7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC(=O)OCC1=C(N2C(C(C2=O)N)SC1)C(=O)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point >300°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 429
Specific Rotation + 94
Solubility Information Solubility in water: soluble
Formula Weight 272.27
Percent Purity 95-102%
Physical Form Crystalline Powder
Chemical Name or Material 7-Aminocephalosporanic acid, 95-102%
Show More Show Less
compliance-icons
Product Identifier
  • 7-Aminocephalosporanic acid
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.