missing translation for 'onlineSavingsMsg'
Learn More

7-Aminocephalosporanic Acid, TRC

Product Code. 30136378
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136378 50 g 50g
17952785 1 g 1g
17920435 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136378 Supplier Toronto Research Chemicals Supplier No. TRCA60341550G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 957-68-6
Molecular Formula C10 H12 N2 O5 S
Molecular Weight (g/mol) 272.28
Synonym (6R,7R)-3-[(Acetyloxy)methyl]-7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, Cefoperazone Sodium Imp. E (EP), Cefazolin Sodium Imp. H (EP), 7-ACA
IUPAC Name (6R,7R)-3-(acetyloxymethyl)-7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC(=O)OCC1=C(N2[C@H](SC1)[C@H](N)C2=O)C(=O)O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C10H12N2O5S/c1-4(13)17-2-5-3-18-9-6(11)8(14)12(9)7(5)10(15)16/h6,9H,2-3,11H2,1H3,(H,15,16)/t6-,9-/m1/s1
Formula Weight 272.0467
Chemical Name or Material (6R,7R)-3-[(Acetyloxy)methyl]-7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.