missing translation for 'onlineSavingsMsg'
Learn More

7-Aminoquinoline, 97%

Product Code. 15516006 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15516006 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15516006 Supplier Thermo Scientific Acros Supplier No. 460050010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 580-19-8 | C9H8N2 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 580-19-8
Melting Point 80.0°C to 86.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C9H8N2
Quantity 1 g
Synonym 7-aminoquinoline, 7-quinolinamine, 7-quinolylamine, quinoline, 7-amino, 7-amino-quinoline, ccris 1682, 7-quinolinamine 9ci, 7-aminequinolin, 7-aminequinoline, 7-quinolinamine acetate
InChI Key RZAUIOKDXQWSQE-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C(C=C2)N)N=C1
IUPAC Name quinolin-7-amine
Molecular Weight (g/mol) 144.17
PubChem CID 11377
Formula Weight 144.17
Percent Purity 97%
Chemical Name or Material 7-Aminoquinoline
Show More Show Less
compliance-icons
Product Identifier
  • 7-Aminoquinoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.